Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GNG2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001243773.1 | NP_001230702.1 | 3, 4 | 632 | ||
NM_001243774.1 | NP_001230703.1 | 2, 3 | 595 | |||
NM_053064.4 | NP_444292.1 | 3, 4 | 636 | |||
XM_006720173.2 | XP_006720236.1 | 3, 4 | 397 | |||
XM_011536846.2 | XP_011535148.1 | 3, 4 | 174 | |||
XM_017021376.1 | XP_016876865.1 | 4, 5 | 418 | |||
XM_017021377.1 | XP_016876866.1 | 5, 6 | 875 | |||
GenBank mRNA | AF493870.1 | - | 1, 2 | 79 | ||
AK001024.1 | - | 3, 4 | 294 | |||
AK092371.1 | - | 2, 3 | 485 | |||
AL832878.1 | - | 3, 4 | 271 | |||
BC020774.1 | - | 3, 4 | 208 | 4243482 | ||
BC060856.1 | - | 2, 3 | 174 | 5276390 | ||
BE245271.1 | - | 2, 3 | 288 | |||
DA226565.1 | - | 3, 4 | 202 | |||
DA785895.1 | - | 3, 4 | 201 |