Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GDAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040875.2 | NP_001035808.1 | 3 | 423 | ||
NM_018972.2 | NP_061845.2 | 3 | 553 | |||
NR_046346.1 | 3 | 408 | ||||
XM_011517552.2 | XP_011515854.1 | 2 | 348 | |||
XM_017013585.1 | XP_016869074.1 | 3 | 1685 | |||
XM_017013586.1 | XP_016869075.1 | 3 | 1685 | |||
GenBank mRNA | AB551556.1 | - | 3 | 552 | ||
AB551557.1 | - | 3 | 552 | |||
AB551558.1 | - | 3 | 550 | |||
AK295594.1 | - | 3 | 347 | |||
BC024939.1 | - | 3 | 388 | 3923335 | ||
BC035450.1 | - | 3 | 400 | |||
Y17849.1 | - | 3 | 481 |