Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285878.1 | NP_001272807.1 | 1 | 2023 | ||
NM_001285879.1 | NP_001272808.1 | 1 | 2023 | |||
NM_005194.3 | NP_005185.2 | 1 | 2023 | |||
GenBank mRNA | AF289608.1 | - | 1 | 1765 | ||
BC005132.2 | - | 1 | 950 | |||
BC007538.1 | - | 1 | 1752 | 3028673 | ||
BC021931.1 | - | 1 | 1767 | 4870789 |