Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WNT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_030761.4 | NP_110388.2 | 5 | 920 | ||
XM_011541597.2 | XP_011539899.1 | 5 | 2394 | |||
XM_011541598.2 | XP_011539900.1 | 5 | 989 | |||
GenBank mRNA | AB061675.1 | - | 5 | 821 | ||
AF316543.1 | - | 5 | 860 | |||
AK296058.1 | - | 5 | 900 | |||
AY358947.1 | - | 5 | 852 | |||
BC057781.1 | - | 5 | 1011 | 30330746 | ||
BT020125.1 | - | 5 | 816 |