Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TERF2IP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018975.3 | NP_061848.2 | 3 | 1110 | ||
GenBank mRNA | AF250393.1 | - | 3 | 1016 | ||
AF262988.1 | - | 3 | 964 | |||
AF289599.1 | - | 3 | 1018 | |||
AK000669.1 | - | 3 | 1102 | |||
AK298880.1 | - | 3 | 808 | |||
AK307901.1 | - | 4 | 1087 | |||
BC004465.1 | - | 3 | 1046 | 2820760 | ||
BC005841.2 | - | 3 | 1046 | 2820760 | ||
BC022428.1 | - | 3 | 1061 | 4253277 | ||
BC078171.1 | - | 3 | 1023 | 5760351 |