Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CEBPD | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_005195.3 | NP_005186.2 | 1 | 301 | ||
GenBank mRNA | BC094715.1 | - | 1 | 553 | ||
BC105109.1 | - | 1 | 296 | 8144112 | ||
M83667.1 | - | 1 | 345 |