Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SSH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161330.1 | NP_001154802.1 | 12 | 1146 | ||
NM_001161331.1 | NP_001154803.1 | 11 | 1238 | |||
NM_018984.3 | NP_061857.3 | 12 | 1146 | |||
XM_005268984.4 | XP_005269041.1 | 12 | 1857 | |||
XM_005268985.2 | XP_005269042.1 | 11 | 972 | |||
XM_011538497.1 | XP_011536799.1 | 11 | 1238 | |||
XM_011538499.1 | XP_011536801.1 | 11 | 1238 | |||
XM_011538500.2 | XP_011536802.1 | 11 | 1062 | |||
XM_011538501.2 | XP_011536803.1 | 11 | 1058 | |||
XM_017019491.1 | XP_016874980.1 | 11 | 2084 | |||
XM_017019492.1 | XP_016874981.1 | 12 | 1199 | |||
XM_017019493.1 | XP_016874982.1 | 10 | 943 | |||
XM_017019494.1 | XP_016874983.1 | 10 | 1038 | |||
GenBank mRNA | AB037719.1 | - | 2 | 181 | ||
AB072355.1 | - | 12 | 1059 | |||
AB072356.1 | - | 12 | 1059 | |||
AK095421.1 | - | 11 | 1239 | |||
BC062341.1 | - | 12 | 1138 | 6376684 |