Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SH3TC1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318480.1 | NP_001305409.1 | 14 | 3431 | ||
NM_018986.4 | NP_061859.3 | 14 | 3348 | |||
NR_134639.1 | 12 | 3062 | ||||
XM_006713889.2 | XP_006713952.1 | 11 | 2766 | |||
XM_006713891.3 | XP_006713954.1 | 8 | 2408 | |||
XM_011513488.1 | XP_011511790.1 | 14 | 3325 | |||
XM_011513489.2 | XP_011511791.1 | 12 | 2948 | |||
XM_017008300.1 | XP_016863789.1 | 14 | 3457 | |||
XM_017008301.1 | XP_016863790.1 | 14 | 3140 | |||
XM_017008302.1 | XP_016863791.1 | 11 | 2850 | |||
GenBank mRNA | AK000363.1 | - | 10 | 2736 | ||
AK074093.1 | - | 13 | 4142 | |||
AK074402.1 | - | 14 | 3225 | |||
AK093562.1 | - | 3 | 1269 | |||
AK126165.1 | - | 14 | 3304 | |||
AK128258.1 | - | 2 | 1042 | |||
AK222865.1 | - | 10 | 2736 | |||
AK310552.1 | - | 3 | 289 |