Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GFOD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242629.1 | NP_001229558.1 | 1 | 988 | ||
NM_018988.3 | NP_061861.1 | 1 | 988 | |||
GenBank mRNA | AK000337.1 | - | 1 | 906 | ||
BC119005.1 | - | 1 | 328 | 40085551 | ||
BC124557.1 | - | 1 | 328 | 40085552 | ||
BC148295.2 | - | 1 | 335 | 40122213 | ||
DA433106.1 | - | 1 | 179 |