Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RIN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242581.1 | NP_001229510.1 | 12 | 2928 | ||
NM_018993.3 | NP_061866.1 | 11 | 2668 | |||
XM_005260731.2 | XP_005260788.1 | 13 | 3124 | |||
XM_005260733.2 | XP_005260790.1 | 9 | 2476 | |||
XM_006723574.3 | XP_006723637.1 | 14 | 3106 | |||
XM_006723575.3 | XP_006723638.1 | 13 | 2892 | |||
XM_006723577.2 | XP_006723640.1 | 12 | 2741 | |||
XM_011529255.2 | XP_011527557.2 | 12 | 3201 | |||
XM_011529257.1 | XP_011527559.1 | 12 | 2797 | |||
XM_011529258.2 | XP_011527560.1 | 12 | 2809 | |||
XM_011529259.2 | XP_011527561.1 | 11 | 2721 | |||
XM_017027887.1 | XP_016883376.1 | 15 | 3447 | |||
XM_017027888.1 | XP_016883377.1 | 16 | 3519 | |||
XM_017027889.1 | XP_016883378.1 | 11 | 3048 | |||
XM_017027890.1 | XP_016883379.1 | 13 | 2879 | |||
XM_017027891.1 | XP_016883380.1 | 13 | 2917 | |||
XM_017027892.1 | XP_016883381.1 | 11 | 2732 | |||
GenBank mRNA | AB060339.1 | - | 11 | 2632 | ||
AL136924.1 | - | 10 | 2549 | |||
BC128065.1 | - | 8 | 1369 | 40035872 | ||
M37190.1 | - | 5 | 1360 |