Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MRPS21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018997.3 | NP_061870.1 | 1 | 583 | ||
NM_031901.5 | NP_114107.1 | 2 | 147 | |||
GenBank mRNA | AB049955.1 | - | 2 | 120 | ||
AF182417.1 | - | 1 | 92 | |||
AK289467.1 | - | 2 | 134 | |||
BC004566.2 | - | 2 | 116 | 3633098 | ||
BC006554.2 | - | 1 | 488 | 2819715 | ||
BG037004.1 | - | 1 | 582 | |||
BI755132.1 | - | 1 | 540 | |||
HY161691.1 | - | 2 | 148 | |||
U79258.1 | - | 1 | 504 |