Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FBXW5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018998.3 | NP_061871.1 | 7 | 1373 | ||
XM_005266089.2 | XP_005266146.2 | 7 | 1423 | |||
XM_011518789.1 | XP_011517091.1 | 7 | 1369 | |||
XM_017014812.1 | XP_016870301.1 | 7 | 1339 | |||
GenBank mRNA | AB209726.1 | - | 4 | 785 | ||
AF217998.1 | - | 3 | 533 | |||
AF258569.1 | - | 3 | 531 | |||
AK026081.1 | - | 7 | 1237 | |||
AK315738.1 | - | 7 | 1248 | |||
AL137631.1 | - | 3 | 927 | |||
BC004541.2 | - | 4 | 782 | |||
BC009429.1 | - | 2 | 526 | |||
BC014130.1 | - | 7 | 1269 | 4620371 | ||
BC014297.2 | - | 7 | 1240 | 3958395 | ||
BC031100.1 | - | 6 | 1334 | |||
BC035355.1 | - | 6 | 1387 |