Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCSER2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001284240.1 | NP_001271169.1 | 6 | 2214 | ||
NM_001284242.1 | NP_001271171.1 | 3 | 295 | |||
NM_001284243.1 | NP_001271172.1 | 3 | 360 | |||
NM_018999.3 | NP_061872.2 | 6 | 2214 | |||
XM_005269905.3 | XP_005269962.1 | 6 | 2245 | |||
XM_005269906.3 | XP_005269963.1 | 6 | 2245 | |||
XM_005269908.3 | XP_005269965.1 | 3 | 348 | |||
XM_006717894.1 | XP_006717957.1 | 4 | 436 | |||
XM_006717895.1 | XP_006717958.1 | 4 | 394 | |||
XM_011539870.2 | XP_011538172.1 | 6 | 2245 | |||
XM_011539871.2 | XP_011538173.1 | 6 | 2245 | |||
XM_011539873.1 | XP_011538175.1 | 4 | 381 | |||
XM_017016340.1 | XP_016871829.1 | 6 | 2134 | |||
XM_017016343.1 | XP_016871832.1 | 4 | 1884 | |||
GenBank mRNA | AB032954.1 | - | 5 | 1217 | ||
AF241785.1 | - | 5 | 1703 | |||
AK024324.1 | - | 5 | 1938 | |||
AK294321.1 | - | 3 | 360 | |||
AK298966.1 | - | 6 | 2140 | |||
BQ440427.1 | - | 3 | 295 |