Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM134B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001034850.2 | NP_001030022.1 | 8 | 1023 | ||
NM_019000.4 | NP_061873.2 | 6 | 899 | |||
XM_011514053.2 | XP_011512355.1 | 9 | 1082 | |||
XM_011514054.2 | XP_011512356.1 | 7 | 1033 | |||
XM_011514055.2 | XP_011512357.1 | 6 | 823 | |||
GenBank mRNA | AK024920.1 | - | 6 | 729 | ||
AK025808.1 | - | 8 | 950 | |||
AL832438.1 | - | 8 | 699 | |||
BC030517.1 | - | 2 | 95 | |||
BC053326.1 | - | 8 | 947 | 5014868 | ||
BC073132.1 | - | 6 | 732 | 5760145 | ||
CR994722.1 | - | 2 | 138 |