Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ANKIB1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019004.1 | NP_061877.1 | 16 | 2522 | ||
XM_005250458.3 | XP_005250515.1 | 16 | 2540 | |||
XM_011516334.1 | XP_011514636.1 | 15 | 2357 | |||
XM_011516335.2 | XP_011514637.1 | 16 | 2104 | |||
XM_017012359.1 | XP_016867848.1 | 16 | 2540 | |||
XM_017012360.1 | XP_016867849.1 | 12 | 1315 | |||
GenBank mRNA | AB037807.1 | - | 16 | 2522 | ||
AK001179.1 | - | 5 | 529 | |||
AK025475.1 | - | 13 | 1619 | |||
AL137349.1 | - | 5 | 504 | |||
BC031483.1 | - | 9 | 1051 | |||
BC063861.1 | - | 16 | 2498 | |||
BC073893.1 | - | 10 | 1114 | |||
BC094865.1 | - | 2 | 204 |