Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TOLLIP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318512.1 | NP_001305441.1 | 5 | 1688 | ||
NM_001318514.1 | NP_001305443.1 | 6 | 1848 | |||
NM_001318515.1 | NP_001305444.1 | 3 | 1411 | |||
NM_001318516.1 | NP_001305445.1 | 5 | 1655 | |||
NM_019009.3 | NP_061882.2 | 6 | 1838 | |||
XM_017017931.1 | XP_016873420.1 | 4 | 1745 | |||
GenBank mRNA | AB209541.1 | - | 7 | 1543 | ||
AK022871.1 | - | 6 | 1745 | |||
AK090850.1 | - | 5 | 1649 | |||
AK096693.1 | - | 5 | 1613 | |||
AK127123.1 | - | 6 | 1802 | |||
AL136835.1 | - | 6 | 1785 | |||
BX648136.1 | - | 1 | 2115 |