Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001042426.1 | NP_001035891.1 | 4 | 812 | ||
NM_001809.3 | NP_001800.1 | 5 | 891 | |||
GenBank mRNA | BC000881.1 | - | 4 | 759 | 3461992 | |
U14518.1 | - | 5 | 851 |