Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190326.1 | NP_001177255.1 | 7 | 2018 | ||
NM_022720.6 | NP_073557.3 | 7 | 2018 | |||
XM_006724268.2 | XP_006724331.1 | 4 | 653 | |||
GenBank mRNA | AK025539.1 | - | 6 | 982 | ||
AK025780.1 | - | 2 | 294 | |||
AK222796.1 | - | 6 | 982 | |||
AY189282.1 | - | 3 | 312 | |||
BC009323.2 | - | 5 | 821 | |||
BC037564.1 | - | 7 | 1968 | |||
BC078147.1 | - | 6 | 1732 | 6014346 | ||
BX649187.1 | - | 4 | 910 | |||
CR456356.1 | - | 6 | 1676 |