Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DGCR8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190326.1 | NP_001177255.1 | 2 | 1090 | ||
NM_022720.6 | NP_073557.3 | 2 | 1090 | |||
GenBank mRNA | AF165527.1 | - | 1 | 751 | ||
AK025539.1 | - | 1 | 54 | |||
AK222796.1 | - | 1 | 54 | |||
AK313357.1 | - | 2 | 1041 | |||
BC009984.2 | - | 2 | 1011 | |||
BC037564.1 | - | 2 | 1040 | |||
BC078147.1 | - | 1 | 804 | 6014346 | ||
CR456356.1 | - | 1 | 748 |