Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMX3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019022.3 | NP_061895.3 | 2, 3 | 244 | ||
XM_005266714.2 | XP_005266771.1 | 2, 3 | 235 | |||
XM_011526033.2 | XP_011524335.1 | 2, 3 | 234 | |||
XM_011526034.2 | XP_011524336.1 | 2, 3 | 236 | |||
XM_011526035.2 | XP_011524337.1 | 2, 3 | 236 | |||
GenBank mRNA | AB058733.1 | - | 2, 3 | 206 | ||
AK122715.1 | - | 2, 3 | 236 | |||
AK295551.1 | - | 2, 3 | 185 | |||
AK307269.1 | - | 2, 3 | 163 | |||
AK310389.1 | - | 2, 3 | 426 | |||
AL832341.1 | - | 2, 3 | 192 | |||
AL832356.1 | - | 2, 3 | 192 | |||
BC007690.2 | - | 1, 2 | 35 | |||
BC018981.2 | - | 1, 2 | 35 | |||
BC032325.2 | - | 2, 3 | 197 | |||
BC093792.1 | - | 2, 3 | 204 | 7939637 | ||
BC093794.1 | - | 2, 3 | 205 | 7939639 | ||
BC107422.1 | - | 2, 3 | 240 | |||
BC143400.1 | - | 2, 3 | 205 | |||
BX647846.1 | - | 2, 3 | 253 |