Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256165.1 | NP_001243094.1 | 2 | 473 | ||
NM_019026.4 | NP_061899.2 | 2 | 273 | |||
NR_045818.1 | 2 | 196 | ||||
GenBank mRNA | AB020980.1 | - | 2 | 240 | ||
AF070626.1 | - | 2 | 229 | |||
AF274935.1 | - | 2 | 109 | |||
AF277194.1 | - | 2 | 147 | |||
AK298614.1 | - | 2 | 440 | |||
AK316080.1 | - | 2 | 406 | |||
AK316610.1 | - | 2 | 219 | |||
AY359027.1 | - | 2 | 208 | |||
AY763589.1 | - | 2 | 202 | |||
BC000104.2 | - | 2 | 193 | 3504808 | ||
BG772940.1 | - | 2 | 278 | |||
BP257387.1 | - | 2 | 169 | |||
CN430121.1 | - | 2 | 251 |