Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256164.1 | NP_001243093.1 | 6, 7 | 529 | ||
NM_001256165.1 | NP_001243094.1 | 6, 7 | 831 | |||
NM_019026.4 | NP_061899.2 | 6, 7 | 631 | |||
NR_045818.1 | 6, 7 | 554 | ||||
GenBank mRNA | AB020980.1 | - | 6, 7 | 598 | ||
AF070626.1 | - | 6, 7 | 587 | |||
AK298614.1 | - | 6, 7 | 798 | |||
AK316080.1 | - | 6, 7 | 764 | |||
AK316610.1 | - | 6, 7 | 577 | |||
AY359027.1 | - | 6, 7 | 566 | |||
AY763589.1 | - | 6, 7 | 560 | |||
BC000104.2 | - | 6, 7 | 551 | 3504808 | ||
BG772940.1 | - | 6, 7 | 637 | |||
BP257387.1 | - | 6, 7 | 527 |