Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001256164.1 | NP_001243093.1 | 4, 5 | 325 | ||
NM_001256165.1 | NP_001243094.1 | 4, 5 | 627 | |||
NM_019026.4 | NP_061899.2 | 4, 5 | 427 | |||
NR_045818.1 | 4, 5 | 350 | ||||
GenBank mRNA | AB020980.1 | - | 4, 5 | 394 | ||
AF070626.1 | - | 4, 5 | 383 | |||
AF274935.1 | - | 3, 4 | 206 | |||
AF277194.1 | - | 4, 5 | 301 | |||
AK298614.1 | - | 4, 5 | 594 | |||
AK316080.1 | - | 4, 5 | 560 | |||
AK316610.1 | - | 4, 5 | 373 | |||
AY359027.1 | - | 4, 5 | 362 | |||
AY763589.1 | - | 4, 5 | 356 | |||
BC000104.2 | - | 4, 5 | 347 | 3504808 | ||
BG772940.1 | - | 4, 5 | 433 | |||
BP257387.1 | - | 4, 5 | 323 | |||
CN430121.1 | - | 4, 5 | 405 |