Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAMTSL4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001288607.1 | NP_001275536.1 | 11, 12 | 2106 | ||
NM_001288608.1 | NP_001275537.1 | 11, 12 | 2106 | |||
NM_019032.5 | NP_061905.2 | 10, 11 | 2037 | |||
NM_025008.4 | NP_079284.2 | 10, 11 | 2037 | |||
XM_011509644.2 | XP_011507946.1 | 10, 11 | 2068 | |||
XM_011509645.2 | XP_011507947.1 | 9, 10 | 1996 | |||
XM_011509648.2 | XP_011507950.1 | 10, 11 | 2022 | |||
XM_011509649.2 | XP_011507951.1 | 10, 11 | 2076 | |||
XM_011509650.2 | XP_011507952.1 | 10, 11 | 2076 | |||
XM_011509651.2 | XP_011507953.1 | 4, 5 | 818 | |||
XM_011509652.1 | XP_011507954.1 | 4, 5 | 524 | |||
XM_017001506.1 | XP_016856995.1 | 11, 12 | 2477 | |||
GenBank mRNA | AF370411.1 | - | 4, 5 | 573 | ||
AK092509.1 | - | 1, 2 | 157 | |||
AY358122.1 | - | 9, 10 | 1851 | |||
BC027478.1 | - | 3, 4 | 476 | |||
BC071852.1 | - | 2, 3 | 333 | |||
BC094811.1 | - | 2, 3 | 333 | |||
BC140800.1 | - | 10, 11 | 1958 | 9021668 | ||
BC144581.1 | - | 10, 11 | 1958 | 9053111 |