Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RHOF | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019034.2 | NP_061907.2 | 5 | 1325 | ||
GenBank mRNA | AK000254.1 | - | 5 | 1291 |