Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142549.1 | NP_001136021.1 | 16 | 1347 | ||
NM_001166533.1 | NP_001160005.1 | 16 | 1389 | |||
NM_001166534.1 | NP_001160006.1 | 10 | 948 | |||
NM_024415.2 | NP_077726.1 | 17 | 1449 | |||
XM_011543495.2 | XP_011541797.1 | 16 | 1394 | |||
XM_011543497.2 | XP_011541799.1 | 15 | 1334 | |||
GenBank mRNA | AF262962.1 | - | 16 | 1367 | ||
AK093439.1 | - | 10 | 922 | |||
AK292417.1 | - | 17 | 1424 | |||
AK308300.1 | - | 16 | 1366 | |||
AL137462.1 | - | 13 | 1085 | |||
AY004154.1 | - | 16 | 1363 | |||
BC047455.1 | - | 16 | 1285 | 5298807 | ||
BC088362.1 | - | 17 | 1408 | 30386875 |