Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CCDC93 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019044.4 | NP_061917.3 | 5, 6 | 639 | ||
XM_006712600.2 | XP_006712663.1 | 5, 6 | 639 | |||
XM_011511359.1 | XP_011509661.1 | 5, 6 | 639 | |||
XM_011511361.1 | XP_011509663.1 | 4, 5 | 393 | |||
XM_011511362.1 | XP_011509664.1 | 4, 5 | 507 | |||
XM_011511363.2 | XP_011509665.1 | 5, 6 | 639 | |||
GenBank mRNA | AB209993.1 | - | 5, 6 | 558 | ||
AK025611.1 | - | 4, 5 | 407 | |||
AK057926.1 | - | 5, 6 | 500 | |||
AK290722.1 | - | 5, 6 | 546 | |||
AK309040.1 | - | 5, 6 | 546 | |||
BC028609.1 | - | 5, 6 | 612 | 4826022 | ||
BC093018.1 | - | 5, 6 | 529 | 30337330 |