Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184965.1 | NP_001171894.1 | 17 | 2776 | ||
NM_001184966.1 | NP_001171895.1 | 15 | 2509 | |||
NM_019045.4 | NP_061918.3 | 17 | 2776 | |||
XM_011531353.2 | XP_011529655.1 | 16 | 2777 | |||
XM_017029600.1 | XP_016885089.1 | 16 | 2709 | |||
GenBank mRNA | AK001978.1 | - | 11 | 1238 | ||
AK092077.1 | - | 14 | 1780 | |||
AK127556.1 | - | 17 | 2687 | |||
AK299711.1 | - | 15 | 2445 | |||
AK304432.1 | - | 17 | 2303 | |||
AL833605.1 | - | 17 | 2741 | |||
AL834210.1 | - | 14 | 2043 | |||
BC028697.2 | - | 17 | 2712 | 4837674 | ||
BX537578.1 | - | 17 | 2750 | |||
BX538087.1 | - | 17 | 2718 | |||
BX648150.1 | - | 17 | 2715 |