Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184965.1 | NP_001171894.1 | 13 | 2200 | ||
NM_001184966.1 | NP_001171895.1 | 12 | 2125 | |||
NM_019045.4 | NP_061918.3 | 13 | 2200 | |||
XM_011531353.2 | XP_011529655.1 | 12 | 2201 | |||
XM_017029600.1 | XP_016885089.1 | 12 | 2133 | |||
GenBank mRNA | AK001978.1 | - | 7 | 668 | ||
AK092077.1 | - | 10 | 1204 | |||
AK127556.1 | - | 13 | 2111 | |||
AK299711.1 | - | 12 | 2061 | |||
AK304432.1 | - | 13 | 1727 | |||
AL833605.1 | - | 13 | 2165 | |||
BC028697.2 | - | 13 | 2136 | 4837674 | ||
BX538087.1 | - | 13 | 2142 | |||
BX648150.1 | - | 13 | 2139 |