Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001184965.1 | NP_001171894.1 | 5 | 1313 | ||
NM_001184966.1 | NP_001171895.1 | 4 | 1238 | |||
NM_019045.4 | NP_061918.3 | 5 | 1313 | |||
XM_011531353.2 | XP_011529655.1 | 4 | 1314 | |||
XM_017029600.1 | XP_016885089.1 | 4 | 1246 | |||
GenBank mRNA | AK092077.1 | - | 2 | 317 | ||
AK127556.1 | - | 5 | 1307 | |||
AK299711.1 | - | 4 | 1174 | |||
AK304432.1 | - | 5 | 840 | |||
AL833605.1 | - | 5 | 1278 | |||
AL834210.1 | - | 2 | 580 | |||
BX537578.1 | - | 5 | 1288 | |||
BX538087.1 | - | 5 | 1255 | |||
BX648150.1 | - | 5 | 1252 |