Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DENND1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001195215.1 | NP_001182144.1 | 22 | 2052 | ||
NR_125340.1 | 19 | 1799 | ||||
XM_005244931.2 | XP_005244988.1 | 21 | 1992 | |||
XM_006711192.2 | XP_006711255.1 | 23 | 1962 | |||
XM_006711193.3 | XP_006711256.1 | 23 | 2094 | |||
XM_006711194.3 | XP_006711257.1 | 10 | 12090 | |||
XM_011509246.2 | XP_011507548.1 | 22 | 1913 | |||
XM_011509247.1 | XP_011507549.1 | 21 | 1853 | |||
XM_011509248.2 | XP_011507550.1 | 22 | 1846 | |||
XM_011509249.2 | XP_011507551.1 | 23 | 2151 | |||
XM_017000470.1 | XP_016855959.1 | 23 | 2040 | |||
XM_017000471.1 | XP_016855960.1 | 22 | 1902 | |||
XM_017000472.1 | XP_016855961.1 | 18 | 1526 | |||
GenBank mRNA | AK000061.1 | - | 8 | 589 | ||
AK091207.1 | - | 19 | 1613 | |||
BC074735.2 | - | 5 | 438 |