Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CENPE | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286734.1 | NP_001273663.1 | 10 | 861 | ||
NM_001813.2 | NP_001804.2 | 10 | 861 | |||
XM_011531544.2 | XP_011529846.1 | 10 | 854 | |||
XM_011531545.2 | XP_011529847.1 | 10 | 860 | |||
XM_011531546.2 | XP_011529848.1 | 10 | 860 | |||
XM_011531547.2 | XP_011529849.1 | 10 | 860 | |||
XM_011531548.2 | XP_011529850.1 | 10 | 860 | |||
XM_011531549.2 | XP_011529851.1 | 10 | 860 | |||
XM_017007659.1 | XP_016863148.1 | 10 | 860 | |||
GenBank mRNA | AB209996.1 | - | 10 | 786 | ||
BQ434767.1 | - | 5 | 277 | |||
Z15005.1 | - | 10 | 861 |