Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CCHCR1 SIRNA Assays ›
Gene Symbol:
CCHCR1
Gene Name:
coiled-coil alpha-helical rod protein 1
Gene Aliases:
C6orf18, HCR, SBP
Chromosome Location:
Chr.6: 31142439 - 31158238 on Build GRCh38
UniGene ID:
Hs.485075
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29153
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CCHCR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001105563.1
NP_001099033.1
5
914
NM_001105564.1
NP_001099034.1
5
1022
NM_019052.3
NP_061925.2
5
689
XM_011514702.2
XP_011513004.1
5
948
XM_011514703.1
XP_011513005.1
6
761
XM_011514704.2
XP_011513006.1
5
550
XM_011514705.1
XP_011513007.1
5
659
XM_017010961.1
XP_016866450.1
5
1056
XM_017010962.1
XP_016866451.1
6
808
XM_017010963.1
XP_016866452.1
6
922
XM_017010964.1
XP_016866453.1
5
686
XM_017010965.1
XP_016866454.1
5
748
XM_017010966.1
XP_016866455.1
5
769
XM_017010967.1
XP_016866456.1
5
872
XM_017010968.1
XP_016866457.1
5
641
XM_017010969.1
XP_016866458.1
5
548
XM_017010970.1
XP_016866459.1
4
334
GenBank mRNA
AB029331.1
-
3
510
AB112474.1
-
5
645
AB112475.1
-
5
996
AF216493.1
-
3
510
AK000204.1
-
5
645
AK000217.1
-
5
646
AK289446.1
-
5
637
AK295494.1
-
5
709
AK310626.1
-
5
707
AY029160.1
-
4
566
BC110534.1
-
5
732
40035162
BC110535.1
-
5
845
40035164
CR457227.1
-
4
566
Back To Top
More Information
Gene Ontology Categories:
Function(s)
protein export from nucleus
multicellular organism development
cell differentiation
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wjc8w:80/10.239.165.44:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0