Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAM35A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019054.2 | NP_061927.2 | 3 | 1540 | ||
XM_006717903.3 | XP_006717966.1 | 3 | 1530 | |||
XM_006717905.3 | XP_006717968.1 | 2 | 1453 | |||
XM_011539876.2 | XP_011538178.1 | 1 | 1431 | |||
XM_011539877.2 | XP_011538179.1 | 4 | 1595 | |||
XM_011539878.2 | XP_011538180.1 | 2 | 1453 | |||
XM_011539879.2 | XP_011538181.1 | 1 | 1439 | |||
XM_011539881.2 | XP_011538183.1 | 1 | 1431 | |||
XM_011539882.2 | XP_011538184.1 | 1 | 1431 | |||
XM_017016349.1 | XP_016871838.1 | 6 | 1839 | |||
XM_017016350.1 | XP_016871839.1 | 3 | 1798 | |||
XM_017016351.1 | XP_016871840.1 | 3 | 1530 | |||
XM_017016352.1 | XP_016871841.1 | 3 | 1530 | |||
GenBank mRNA | AK022978.1 | - | 3 | 1540 | ||
AK025606.1 | - | 1 | 389 | |||
BC051863.2 | - | 2 | 1486 | 6146593 |