Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDIT4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019058.2 | NP_061931.1 | 3 | 1411 | ||
GenBank mRNA | AF335324.1 | - | 3 | 1427 | ||
AK000507.1 | - | 3 | 1406 | |||
AL136668.1 | - | 3 | 1410 | |||
AY090097.1 | - | 3 | 1327 | |||
BC000708.1 | - | 3 | 1317 | 3507244 | ||
BC007714.1 | - | 3 | 1399 | 4302878 | ||
BC015236.1 | - | 3 | 1385 | 3906374 | ||
BM843387.1 | - | 1 | 251 | |||
CN428398.1 | - | 1 | 27 |