Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EGLN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022051.2 | NP_071334.1 | 4 | 4354 | ||
XM_005273167.4 | XP_005273224.1 | 3 | 1503 | |||
GenBank mRNA | AF229245.1 | - | 4 | 4354 | ||
AF277174.1 | - | 4 | 312 | |||
AJ310543.1 | - | 4 | 1294 | |||
AL833885.1 | - | 4 | 722 | |||
BC005369.1 | - | 4 | 386 | |||
DQ975380.1 | - | 4 | 443 |
Gene Symbol: SCAND2P | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AF229246.1 | - | 2 | 1598 | ||
AL109690.1 | - | 1 | 338 |