Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DDX56 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001257189.1 | NP_001244118.1 | 8 | 1676 | ||
NM_019082.3 | NP_061955.1 | 9 | 1796 | |||
GenBank mRNA | AF247666.1 | - | 9 | 1186 | ||
AJ131712.1 | - | 9 | 1220 | |||
AK022774.1 | - | 9 | 1167 | |||
AK125595.1 | - | 9 | 2342 | |||
AK222662.1 | - | 9 | 1185 | |||
AK309276.1 | - | 8 | 1036 | |||
AK315363.1 | - | 8 | 1065 | |||
AL136700.1 | - | 9 | 1162 | |||
BC001235.1 | - | 9 | 1183 | 3456547 | ||
CR457213.1 | - | 9 | 1153 |