Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ARL15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019087.2 | NP_061960.1 | 4 | 495 | ||
XM_011543498.2 | XP_011541800.1 | 6 | 600 | |||
XM_011543499.2 | XP_011541801.1 | 5 | 543 | |||
XM_011543500.2 | XP_011541802.1 | 5 | 569 | |||
XM_017009598.1 | XP_016865087.1 | 6 | 587 | |||
GenBank mRNA | AK000058.1 | - | 4 | 495 | ||
AK290886.1 | - | 4 | 496 | |||
AK311482.1 | - | 4 | 418 | |||
BC026093.1 | - | 4 | 499 | 4699068 | ||
CR457225.1 | - | 4 | 401 |