Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: HES2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019089.4 | NP_061962.2 | 4 | 916 | ||
GenBank mRNA | AK023754.1 | - | 1 | 101 | ||
AK091122.1 | - | 4 | 916 | |||
BC132698.1 | - | 4 | 879 | 40146720 | ||
BC136963.1 | - | 4 | 879 | 9020955 | ||
BC142687.1 | - | 4 | 896 | 40148619 |