Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UGT1A3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019093.2 | NP_061966.1 | 1 | 592 | ||
GenBank mRNA | AY435138.1 | - | 1 | 592 |