Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TBC1D13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_018201.4 | NP_060671.3 | 9 | 928 | ||
XM_005252060.2 | XP_005252117.1 | 8 | 831 | |||
GenBank mRNA | AB209565.1 | - | 6 | 647 | ||
AB449899.1 | - | 9 | 781 | |||
AK123828.1 | - | 5 | 542 | |||
AK289777.1 | - | 9 | 840 | |||
AK295183.1 | - | 6 | 631 | |||
AK307556.1 | - | 5 | 530 | |||
BC014898.1 | - | 9 | 803 | |||
BC132817.1 | - | 9 | 801 | 40146839 | ||
BC136509.1 | - | 9 | 801 | 9020499 | ||
BC150311.1 | - | 9 | 837 | 8860447 |