Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: WDR74 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001307977.1 | NP_001294906.1 | 6, 7 | 712 | ||
NM_018093.3 | NP_060563.2 | 7, 8 | 791 | |||
XM_005274055.2 | XP_005274112.1 | 7, 8 | 2652 | |||
GenBank mRNA | AK001301.1 | - | 7, 8 | 791 | ||
AK025383.1 | - | 2, 3 | 226 | |||
AK292330.1 | - | 7, 8 | 1161 | |||
AY500828.1 | - | 6, 7 | 643 | |||
BC006351.2 | - | 6, 7 | 634 | 4111690 |