Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMEM106B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134232.1 | NP_001127704.1 | 7, 8 | 887 | ||
NM_018374.3 | NP_060844.2 | 8, 9 | 1081 | |||
XM_005249789.1 | XP_005249846.1 | 7, 8 | 937 | |||
GenBank mRNA | AK002135.1 | - | 7, 8 | 811 | ||
AK223263.1 | - | 7, 8 | 785 | |||
BC028108.1 | - | 7, 8 | 801 | |||
BC033901.2 | - | 8, 9 | 1003 | 5263860 | ||
BC039741.2 | - | 7, 8 | 798 | 6066301 |