Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: GTPBP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286216.1 | NP_001273145.1 | 2 | 90 | ||
NM_019096.4 | NP_061969.3 | 2 | 285 | |||
XM_017010976.1 | XP_016866465.1 | 2 | 279 | |||
GenBank mRNA | AB024574.1 | - | 3 | 128 | ||
AF168990.1 | - | 2 | 82 | |||
AK000430.1 | - | 2 | 47 | |||
AK290267.1 | - | 2 | 90 | |||
AL834331.1 | - | 2 | 67 | |||
BC064968.1 | - | 2 | 238 | 6149902 |