Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143935.1 | NP_001137407.1 | 13 | 1484 | ||
NM_021151.3 | NP_066974.2 | 12 | 1400 | |||
XM_011516337.2 | XP_011514639.1 | 12 | 1400 | |||
XM_017012370.1 | XP_016867859.1 | 13 | 1474 | |||
GenBank mRNA | AF073770.1 | - | 12 | 1291 | ||
AF168793.1 | - | 12 | 1291 | |||
AK127901.1 | - | 13 | 1440 | |||
AK300114.1 | - | 7 | 730 | |||
AK313487.1 | - | 12 | 1343 | |||
BC039004.1 | - | 12 | 1340 | 5242962 |