Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CROT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001143935.1 | NP_001137407.1 | 6 | 610 | ||
NM_021151.3 | NP_066974.2 | 5 | 526 | |||
XM_011516337.2 | XP_011514639.1 | 5 | 526 | |||
XM_017012370.1 | XP_016867859.1 | 5 | 464 | |||
GenBank mRNA | AF073770.1 | - | 5 | 417 | ||
AF168793.1 | - | 5 | 417 | |||
AK127901.1 | - | 6 | 566 | |||
AK313487.1 | - | 5 | 470 | |||
BC039004.1 | - | 5 | 466 | 5242962 | ||
DA518578.1 | - | 5 | 524 | |||
DA865171.1 | - | 5 | 526 |