Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNHIT6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170670.1 | NP_001164141.1 | 2 | 403 | ||
NM_017953.3 | NP_060423.3 | 1 | 520 | |||
XM_011541614.2 | XP_011539916.1 | 1 | 534 | |||
GenBank mRNA | AF308296.1 | - | 1 | 167 | ||
AK000767.1 | - | 1 | 429 | |||
AK298149.1 | - | 2 | 403 | |||
AK314566.1 | - | 1 | 448 | |||
AL442074.1 | - | 1 | 505 | |||
BC026236.1 | - | 1 | 507 | 4664103 | ||
BC110898.1 | - | 1 | 438 | 6012059 | ||
DB169943.1 | - | 1 | 520 |