Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRN3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001301064.1 | NP_001287993.1 | 17 | 1891 | ||
NM_018427.4 | NP_060897.3 | 18 | 1981 | |||
XM_005255376.1 | XP_005255433.1 | 16 | 1756 | |||
XM_005255377.4 | XP_005255434.1 | 15 | 1705 | |||
XM_011522540.2 | XP_011520842.1 | 13 | 1393 | |||
XM_011522541.1 | XP_011520843.1 | 17 | 1820 | |||
GenBank mRNA | AF227156.1 | - | 18 | 1947 | ||
AJ272050.1 | - | 18 | 1901 | |||
AK225662.1 | - | 18 | 1957 | |||
AK295486.1 | - | 15 | 1574 | |||
AK300253.1 | - | 17 | 1800 | |||
AK302994.1 | - | 15 | 1628 | |||
AK303405.1 | - | 14 | 1474 | |||
AK304856.1 | - | 18 | 1847 | |||
AK314769.1 | - | 18 | 1895 | |||
AK316520.1 | - | 17 | 1805 | |||
BC036182.1 | - | 18 | 1896 | |||
BC071868.1 | - | 10 | 1216 | |||
BC132688.1 | - | 18 | 1898 | 40146710 | ||
BC132690.1 | - | 18 | 1898 | 40146712 |