Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161779.1 | NP_001155251.1 | 3 | 1111 | ||
NM_001161780.1 | NP_001155252.1 | 3 | 985 | |||
NM_001161781.1 | NP_001155253.1 | 2 | 972 | |||
NM_018444.3 | NP_060914.2 | 2 | 1029 | |||
XM_011517135.2 | XP_011515437.1 | 3 | 882 | |||
XM_011517136.1 | XP_011515438.1 | 3 | 989 | |||
XM_017013587.1 | XP_016869076.1 | 2 | 1044 | |||
XM_017013588.1 | XP_016869077.1 | 3 | 952 | |||
GenBank mRNA | AF155661.1 | - | 3 | 999 | ||
AK057079.1 | - | 2 | 941 | |||
AK126862.1 | - | 1 | 1406 | |||
AK302651.1 | - | 2 | 915 | |||
BC047619.1 | - | 2 | 950 | 4824168 | ||
BC064978.1 | - | 3 | 969 | |||
BC098343.1 | - | 1 | 783 | 40010016 |