Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other RBFOX1 SIRNA Assays ›
Gene Symbol:
RBFOX1
Gene Name:
RNA binding protein, fox-1 homolog 1
Gene Aliases:
2BP1, A2BP1, FOX-1, FOX1, HRNBP1
Chromosome Location:
Chr.16: 5239749 - 7713343 on Build GRCh38
UniGene ID:
Hs.459842
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s29339
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: RBFOX1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001142333.1
NP_001135805.1
7, 8
1466
NM_001142334.1
NP_001135806.1
5, 6
698
NM_001308117.1
NP_001295046.1
7, 8
803
NM_018723.3
NP_061193.2
7, 8
1466
NM_145891.2
NP_665898.1
4, 5
790
NM_145892.2
NP_665899.1
4, 5
790
NM_145893.2
NP_665900.1
4, 5
790
XM_005255386.3
XP_005255443.1
4, 5
716
XM_005255387.3
XP_005255444.1
4, 5
695
XM_005255388.4
XP_005255445.1
4, 5
751
XM_005255390.3
XP_005255447.1
7, 8
1588
XM_005255391.3
XP_005255448.1
6, 7
598
XM_005255394.4
XP_005255451.1
4, 5
695
XM_011522546.2
XP_011520848.1
4, 5
695
XM_011522547.2
XP_011520849.1
4, 5
695
XM_011522548.2
XP_011520850.1
4, 5
695
XM_017023318.1
XP_016878807.1
8, 9
1093
XM_017023319.1
XP_016878808.1
10, 11
1089
XM_017023320.1
XP_016878809.1
7, 8
1497
XM_017023321.1
XP_016878810.1
6, 7
2559
XM_017023322.1
XP_016878811.1
6, 7
2559
XM_017023323.1
XP_016878812.1
4, 5
716
XM_017023324.1
XP_016878813.1
7, 8
1497
XM_017023325.1
XP_016878814.1
5, 6
1095
XM_017023326.1
XP_016878815.1
7, 8
1588
XM_017023327.1
XP_016878816.1
4, 5
716
XM_017023328.1
XP_016878817.1
7, 8
1497
XM_017023329.1
XP_016878818.1
7, 8
1497
XM_017023330.1
XP_016878819.1
6, 7
2036
XM_017023331.1
XP_016878820.1
4, 5
2502
XM_017023332.1
XP_016878821.1
4, 5
716
XM_017023333.1
XP_016878822.1
6, 7
598
XM_017023334.1
XP_016878823.1
4, 5
716
XM_017023335.1
XP_016878824.1
7, 8
1588
XM_017023336.1
XP_016878825.1
6, 7
598
XM_017023337.1
XP_016878826.1
4, 5
716
XM_017023338.1
XP_016878827.1
6, 7
598
XM_017023339.1
XP_016878828.1
4, 5
742
XM_017023341.1
XP_016878830.1
7, 8
1588
XM_017023342.1
XP_016878831.1
6, 7
598
XM_017023343.1
XP_016878832.1
4, 5
745
GenBank mRNA
AB208826.1
-
1500
AF094849.1
-
4, 5
791
AF107203.1
-
7, 8
1464
AF109106.1
-
4, 5
791
AF229057.1
-
4, 5
793
AF448859.1
-
7, 8
1464
AK001027.1
-
5, 6
698
AK293951.1
-
7, 8
803
AL713700.1
-
6, 7
1313
BC113691.1
-
4, 5
584
8322743
BC143816.1
-
4, 5
584
KJ534926.1
-
4, 5
538
KJ535000.1
-
4, 5
538
KJ535001.1
-
4, 5
538
Back To Top
More Information
Other Species Targeted
This siRNA also targets the following transcripts in Rat: NM_001106974, XM_006245774.This siRNA also targets the following transcripts in Mouse: NM_021477, NM_183188, AY659954, AB041596, AY659955, and others.
Panther Classification:
Molecular Function -
RNA metabolism protein
RNA processing factor
RNA splicing factor
nucleic acid metabolism protein
Gene Ontology Categories:
Function(s)
Process(es)
regulation of alternative mRNA splicing, via spliceosome
mRNA processing
nervous system development
RNA splicing
RNA transport
nucleotide binding
RNA binding
mRNA binding
protein binding
protein C-terminus binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-nnwcq:80/10.239.162.212:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0